Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP5 Recombinant Rabbit Monoclonal Antibody (HL1367)
GREENER_CHOICE

Catalog No. PIMA547017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547017 Supplier Invitrogen™ Supplier No. MA547017
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

SARS-CoV-2 NSP5 is a non-structural protein in which the main protease targets RIG-I and mitochondrial antiviral signaling (MAVS) protein via two distinct mechanisms for inhibition. NSP5 cleaves off the 10 most-N-terminal amino acids from RIG-I and deprives it of the ability to activate MAVS, whereas NSP5 promotes the ubiquitination and proteosome-mediated degradation of MAVS. NSP5 potently inhibits interferon (IFN) induction by double-stranded RNA (dsRNA) in an enzyme-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP5
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1367
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ORF1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols ORF1ab
Host Species Rabbit
Immunogen Recombinant fragment of SARS-CoV-2 (COVID-19) 3CLpro (nsp5).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.