Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-COV-2 NSP5 Rabbit anti-Virus, Polyclonal, ProteinTech
SDP

Catalog No. 502238853 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8853 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8853 Supplier Proteintech Group Inc Supplier No. 292861AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

SARS-COV-2 NSP5 Polyclonal antibody specifically detects SARS-COV-2 NSP5 in Virus samples. It is validated for ELISA, Immunocytochemistry

Specifications

Antigen SARS-COV-2 NSP5
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 450 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ORF1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols ORF1ab
Host Species Rabbit
Immunogen NSP5 Fusion Protein Ag30934
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.