Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143824
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143824 Supplier Invitrogen™ Supplier No. PA5143824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 NSP6 Polyclonal Antibody for ELISA

Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP6
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias CoV; CoV-2; NSP6; SARS
Host Species Rabbit
Immunogen A peptide corresponding to 18 amino acids in the center of SARS-CoV-2 (COVID-19) NSP6 protein. The immunogen is located in between 230-280 amino acids of the SARS-CoV-2 (COVID-19) NSP6 protein.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.