Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120714
This item is not returnable.
View return policy
Description
Positive test controls include: 293T. Immunogen sequence: RLTLGVYDYL VSTQEFRYMN SQGLLPPKNS IDAFKLNIKL LGVGGKPCIK VATVQ.
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.
Specifications
SARS-CoV-2 NSP6 | |
Polyclonal | |
Unconjugated | |
CoV; CoV-2; NSP6; SARS | |
Recombinant fusion protein containing a sequence corresponding to amino acids 236-290 of coronavirus NSP6 (YP_0097253021). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction