Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp8 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307047
Description
SARS-CoV-2 nsp8 Polyclonal specifically detects SARS-CoV-2 nsp8 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA.Specifications
SARS-CoV-2 nsp8 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
43740578 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 0.5 ug/mL, ELISA | |
Non-structural protein 8, nsp8 replicase | |
This antibody was raised against a peptide corresponding to 14 amino acids near the center of SARS-CoV-2 (COVID-19) NSP8 protein. The immunogen is located within 40-90 amino acids of SARS-CoV-2 (COVID-19) NSP8. | |
0.1 mg | |
Primary | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction