Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143469 Supplier Invitrogen™ Supplier No. PA5143469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The nonstructural protein 8 (nsp8) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp8 is thought to be part of the viral replication complex, which is associated with intracellular membranes. No specific information on the function of nsp8 is available.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias CoV-2; non-structural protein 8; NSP8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.