Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp9 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307969
Description
SARS-CoV-2 nsp9 Polyclonal specifically detects SARS-CoV-2 nsp9 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA.Specifications
SARS-CoV-2 nsp9 | |
Polyclonal | |
Western Blot 1:1000-1:5000, ELISA 1:4000-1:8000 | |
Non-structural protein 9, nsp9 replicase | |
Recombinant SARS-CoV-2 NSP9(Asn4141-Gln4253) | |
0.05 mg | |
Primary | |
SARS-CoV-2 | |
Purified |
Western Blot, ELISA | |
Unconjugated | |
PBS, pH 7.4, 50% glycerol. | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
43740578 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction