Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody (040), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312736H
Description
SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
SARS-CoV-2 Nucleocapsid | |
Monoclonal | |
HRP | |
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
Recombinant SARS-CoV-2 Nucleocapsid Protein | |
100 μg | |
Virology Bacteria and Parasites | |
43740575 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
040 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
SARS-CoV | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction