Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody (B3449M), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP300514APC
Description
SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Lateral Flow, ELISA (Matched Antibody Pair)Specifications
SARS-CoV-2 Nucleocapsid | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
SARS-CoV, SARS-CoV-2 | |
Purified |
Western Blot, ELISA, Lateral Flow, ELISA Detection (Matched Antibody Pair) | |
B3449M | |
Western Blot, ELISA, Lateral Flow Assay, ELISA Detection (Matched Antibody Pair) | |
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9, structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
Recombinant SARS-CoV-2 Nucleocapsid Protein. | |
0.2 mL | |
Virology Bacteria and Parasites | |
43740575 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction