Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $487.50
Specifications
Antigen | SARS-CoV-2 Nucleocapsid |
---|---|
Concentration | 1.0 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SARS-CoV-2 Nucleocapsid Polyclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2, Virus samples. It is validated for Western Blot, Western BlotSpecifications
SARS-CoV-2 Nucleocapsid | |
Western Blot | |
Unconjugated | |
RUO | |
SARS-CoV-2, Virus | |
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
Full length recombinant SARS-CoV-2 nucleocapsid protein. | |
Immunogen affinity purified |
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Virology Bacteria and Parasites | |
PBS | |
43740575 | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title