Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300510JF646
This item is not returnable.
View return policy
Description
SARS-CoV-2 Nucleocapsid Polyclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2, Virus samples. It is validated for Western BlotSpecifications
SARS-CoV-2 Nucleocapsid | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Immunogen Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2, Virus | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
Full length recombinant SARS-CoV-2 nucleocapsid protein. | |
0.1 mL | |
Virology Bacteria and Parasites | |
43740575 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction