Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Nucleocapsid |
---|---|
Clone | 2815D |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10857SP
![]() |
R&D Systems
MAB10857SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10857100
![]() |
R&D Systems
MAB10857100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in SARS-CoV-2 samples. It is validated for ELISASpecifications
Nucleocapsid | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived SARS-CoV-2 Nucleocapsid, Met1-Ala419, Accession # YP_009724397.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2815D | |
ELISA | |
Unconjugated | |
Rabbit | |
SARS-CoV-2 | |
YP_009724397.2 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title