Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid Mouse anti-SARS-CoV, SARS-CoV-2, Clone: B3449M, R&D Systems™
SDP

Catalog No. NB092787 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092787 200 μL
NB092786 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092787 Supplier R&D Systems Supplier No. NBP300514200UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SARS-CoV-2 Nucleocapsid Monoclonal specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV/SARS-CoV-2 samples. It is validated for ELISA/ELISA Detection (Matched Antibody Pair)/Lateral Flow Assay/Western Blot.

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications ELISA, ELISA Detection (Matched Antibody Pair), Lateral Flow, Western Blot
Classification Monoclonal
Clone B3449M
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation Phosphate Buffered Saline, pH 7.4
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, Nucleocapsid protein, Nucleoprotein, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Host Species Mouse
Immunogen Recombinant SARS-CoV-2 Nucleocapsid Protein.
Purification Method Protein G purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.