Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid protein Monoclonal Antibody (H4G1), Huabio

Catalog No. 502544528
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4528 Supplier HUABIO RESEARCH INC Supplier No. HA600006
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV-2 Nucleocapsid protein Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid protein in Virus samples. It is validated for ELISA, Western Blot

Specifications

Antigen SARS-CoV-2 Nucleocapsid protein
Applications ELISA, Western Blot
Classification Monoclonal
Clone H4G1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene N
Gene Accession No. P0DTC9
Gene Alias COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
Host Species Mouse
Immunogen Recombinant full length protein corresponding to SARS-CoV-2 nucleocapsid protein aa 1-419.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.