Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Nucleocapsid/Spike Protein (RBD) Recombinant Protein

Catalog No. pirp87706
Encompass
Click to view available options
Quantity:
100 μg

Recombinant Protein

Animal Origin Free Purity is >95% by SDS-PAGE.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.5 mg/mL
For Use With (Application) Control, ELISA
Formulation PBS with no preservative; pH 7.4
Name SARS-CoV-2 Nucleocapsid/Spike Protein (RBD)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein RBD
Conjugate Unconjugated
Recombinant Recombinant
Sequence Full length SARS-CoV-2 Nucleoprotein/Spike Protein (RBD)
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Expression System E. coli
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.