Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Omicron (XBB.1) Spike Antibody (D008), Alexa Fluor™ 750 , Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP318853AF750
Description
SARS-CoV-2 Omicron (XBB.1) Spike Monoclonal antibody specifically detects SARS-CoV-2 Omicron (XBB.1) Spike in SARS-CoV-2 samples. It is validated for ELISASpecifications
SARS-CoV-2 Omicron (XBB.1) Spike | |
Monoclonal | |
Alexa Fluor 750 | |
Human | |
Protein A purified | |
RUO | |
SARS-CoV-2 | |
Purified |
ELISA | |
D008 | |
50mM Sodium Borate | |
This is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Omicron XBB.1 Spike ECD Protein. The antibody was produced using recombinant antibody technology. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction