Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 ORF6 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$160.20 - $389.00
Specifications
Antigen | SARS-CoV-2 ORF6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SARS-CoV-2 ORF6 Polyclonal antibody specifically detects SARS-CoV-2 ORF6 in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
SARS-CoV-2 ORF6 | |
Polyclonal | |
Purified | |
RUO | |
SARS-CoV-2 | |
2019-nCoV ORF6, 2019-nCoV ORF6 Protein, Accessory Protein 6, COVID-19 Non-structural protein 6, COVID-19 ns6, COVID-19 ORF6, COVID-19 Protein X3, Human coronavirus ORF6 Protein, Non-structural protein 6, NS6, ORF6 protein, SARS-CoV-2, SARS-CoV-2 Accessory Protein 6, SARS-CoV-2 Non-structural protein 6, SARS-CoV-2 ns6, SARSCoV2 ORF6 Protein, SARS-CoV-2 ORF6 Protein, SARS-CoV-2 Protein X3, Severe Acute Respiratory Syndrome Coronavirus 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-61 of coronavirus ORF6 (YP_009724394.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Infections (Virus Bacteria and Parasites) | |
PBS, 50% glycerol, pH7.3 | |
43740572 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title