Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 ORF9b Antibody (HL1918) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325670
Description
SARS-CoV-2 ORF9b Monoclonal antibody specifically detects SARS-CoV-2 ORF9b in SARS-CoV-2 samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
SARS-CoV-2 ORF9b | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry | |
HL1918 | |
Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:1000 | |
Accessory protein 9b, ORF9b, ORF-9b, Protein 9b | |
Full length SARS-CoV-2 ORF9b recombinant protein. | |
100 μL | |
Infections (Virus Bacteria and Parasites) | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction