Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 ORF9b Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143485 Supplier Invitrogen™ Supplier No. PA5143485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

ORF9b plays a role in the inhibition of host innate immune response by targeting the mitochondrial-associated adapter MAVS. Mechanistically, usurps the E3 ligase ITCH to trigger the degradation of MAVS, TRAF3, and TRAF6. In addition, causes mitochondrial elongation by triggering ubiquitination and proteasomal degradation of dynamin-like protein 1/DNM1L.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF9b
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias COVID19 ORF9b; COVID-19 ORF9b; ORF9b; SARS-CoV-2 Open Reading Frame 9b; SARS-COV2 ORF9b
Host Species Rabbit
Immunogen Full length SARS-CoV-2 (COVID-19) ORF9b recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.