Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 ORF9b Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116951
Description
Positive Samples: 293T Immunogen sequence: MDPKISEMHP ALRLVDPQIQ LAVTRMENAV GRDQNNVGPK VYPIILRLGS PLSLNMARKT LNSLEDKAFQ LTPIAVQMTK LATTEELPDE FVVVTVK.
ORF9b plays a role in the inhibition of host innate immune response by targeting the mitochondrial-associated adapter MAVS. Mechanistically, usurps the E3 ligase ITCH to trigger the degradation of MAVS, TRAF3, and TRAF6. In addition, causes mitochondrial elongation by triggering ubiquitination and proteasomal degradation of dynamin-like protein 1/DNM1L.
Specifications
SARS-CoV-2 ORF9b | |
Polyclonal | |
Unconjugated | |
COVID19 ORF9b; COVID-19 ORF9b; ORF9b; SARS-CoV-2 Open Reading Frame 9b; SARS-COV2 ORF9b | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-97 of coronavirus ORF9b (P0DTD2). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.55 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction