Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 ORF9b Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $389.00
Specifications
Antigen | SARS-CoV-2 ORF9b |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SARS-CoV-2 ORF9b Polyclonal antibody specifically detects SARS-CoV-2 ORF9b in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
SARS-CoV-2 ORF9b | |
Western Blot | |
Unconjugated | |
Rabbit | |
Infections (Virus Bacteria and Parasites) | |
PBS, 50% glycerol, pH7.3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-97 of coronavirus ORF9b (P0DTD2). MDPKISEMHPALRLVDPQIQLAVTRMENAVGRDQNNVGPKVYPIILRLGSPLSLNMARKTLNSLEDKAFQLTPIAVQMTKLATTEELPDEFVVVTVK | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
SARS-CoV-2 | |
Accessory protein 9b, ORF9b, ORF-9b, Protein 9b | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title