Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 ORF9c Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143834
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143834 Supplier Invitrogen™ Supplier No. PA5143834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF9c Polyclonal Antibody for Western Blot, ELISA

SARS-CoV-2 ORF9c is a protein that is unique to the SARS-CoV-2 virus and is not found in other coronaviruses. Its function is not yet fully understood, but studies suggest that it may be involved in modulating the immune response of the host. ORF9c is believed to interact with the host's immune system, potentially suppressing the production of interferons, which are essential for fighting viral infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF9c
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ORF7a
Gene Alias ORF14; Uncharacterized protein 14
Host Species Rabbit
Immunogen A peptide corresponding to 16 amino acids near the amino-terminus of SARS-CoV-2 (COVID-19) ORF9c protein. The immunogen is located within the first 50 amino acids of the SARS-CoV-2 (COVID-19) ORF9c protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740573
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.