Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 RBD Neutralizing Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
SDP

Catalog No. 502543259
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3259 Supplier OriGene Supplier No. CF814512
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV-2 RBD Neutralizing Monoclonal antibody specifically detects SARS-CoV-2 RBD Neutralizing in Virus samples. It is validated for ELISA, Neutralization

Specifications

Antigen SARS-CoV-2 RBD Neutralizing
Applications ELISA, Neutralization
Classification Monoclonal
Clone OTI1C2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene N
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Host Species Mouse
Immunogen Full length recombinant SARS-CoV-2 N protein produced in HEK293.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.