Learn More
SARS-CoV-2 S protein (944-1214 aa) Rabbit anti-Virus, Polyclonal, ProteinTech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 288671AP150UL
Description
Aliquoting is unnecessary for -20°C storage.
SARS-CoV-2 S protein (944-1214 aa) Polyclonal antibody specifically detects SARS-CoV-2 S protein (944-1214 aa) in Virus samples. It is validated for ELISASpecifications
| SARS-CoV-2 S protein (944-1214 aa) | |
| Polyclonal | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 S2; COVID19 Spike S2; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| spike protein Fusion Protein Ag30685 | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
| ELISA | |
| 375 μg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.