Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike Antibody (8A5) - Azide and BSA Free, Novus Biologicals™

Monkey Monoclonal Antibody
Supplier: Novus Biologicals NBP307034
Description
SARS-CoV-2 Spike Monoclonal specifically detects SARS-CoV-2 Spike in SARS-CoV-2 samples. It is validated for ELISA, Neutralization.Specifications
SARS-CoV-2 Spike | |
Monoclonal | |
2 mg/mL | |
ELISA, Neutralization | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
Cultured in vitro under conditions free from animal derived components. | |
0.025 mg | |
Primary | |
SARS-CoV-2 | |
IgG1 |
ELISA, Neutralization | |
8A5 | |
Unconjugated | |
PBS, pH 7.4 | |
Monkey | |
>95% | |
RUO | |
43740568 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. After opening store under sterile conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction