Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike Antibody (CR3022), DyLight 350 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290979UV
Description
SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.Specifications
SARS-CoV-2 Spike | |
Monoclonal | |
DyLight 350 | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, Human coronavirus spike glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein | |
Rabbit | |
Protein A purified | |
RUO | |
This antibody binds to both SARS-CoV and SARS-CoV-2 with high affinity (PMID: 16796401 & 32065055). It binds the amino acids 318-510 in the S1 domain of the SARS-CoV Spike protein as well as SARS-CoV-2 (COVID-19) Spike protein. The antibody also binds to P462L-substituted S318-510 fragments of the SARS spike protein. The binding epitope is only accessible in the "open" confromation of the spike protein (Joyce et al. 2020). NBP2-90979 cross-reacts with spike protein of Omicron and Delta variants. | |
Store at 4°C in the dark. | |
IgG κ |
ELISA | |
CR3022 | |
50mM Sodium Borate | |
S | |
The original monoclonal antibody was generated by sequencing peripheral blood lymphocytes of a patient exposed to the SARS-CoV. | |
0.1 mL | |
Primary | |
SARS-CoV | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction