Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike Antibody (CR3022), FITC, R&D Systems™
SDP

Catalog No. NB153166 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153166 Supplier R&D Systems Supplier No. NBP312017F

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SARS-CoV-2 Spike Monoclonal specifically detects SARS-CoV-2 Spike in SARS-CoV, SARS-CoV-2 samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Neutralization, Surface Plasmon Resonance, X-Ray Crystallography.

Specifications

Antigen SARS-CoV-2 Spike
Applications ELISA, Immunocytochemistry, Immunofluorescence, Neutralization, Surface Plasmon Resonance, X-Ray Crystallography
Classification Monoclonal
Clone CR3022
Conjugate FITC
Formulation PBS
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Host Species Mouse
Immunogen The original monoclonal antibody was generated by sequencing peripheral blood lymphocytes of a patient exposed to the SARS-CoV.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.