Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike Mouse anti-SARS-CoV-2, SARS-CoV, Clone: CR3022, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | SARS-CoV-2 Spike |
---|---|
Clone | CR3022 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence, Surface Plasmon Resonance, Neutralization, X-Ray Crystallography |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA, Immunofluorescence, Neutralization, X-Ray CrystallographySpecifications
SARS-CoV-2 Spike | |
ELISA, Immunocytochemistry/Immunofluorescence, Surface Plasmon Resonance, Neutralization, X-Ray Crystallography | |
Unconjugated | |
RUO | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
The original monoclonal antibody was generated by sequencing peripheral blood lymphocytes of a patient exposed to the SARS-CoV. | |
Protein A purified |
CR3022 | |
Monoclonal | |
Mouse | |
PBS | |
43740568 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title