Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike (N501Y Mutant) Recombinant Rabbit Monoclonal Antibody (1J5G2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551234
Description
Immunogen sequence: NYLYRLFRKS NLKPFERDIS TEIYQAGSTP CNGVEGFNCY FPLQSYGFQP TYGVGYQPYR VVVLSFELLH APATVCGPKK STNLVKNKCV NFNFNGLTGT G.
The spike protein in SARS-CoV-2 (SARS-2-S) interacts with the human ACE2 receptor to gain entry into a cell to initiate infection SARS-2-S is cleaved into S1 and S2 subunits, with S1 serving the function of receptor-binding and S2 serving the function of membrane fusion The S protein undergoes two crucial cleavage events, with the first splitting S1 and S2 and the second splitting S2 into FP and S2' [PMID: 33466921].
Specifications
SARS-CoV-2 Spike (N501Y Mutant) | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
Liquid |
ELISA, Western Blot | |
1J5G2 | |
Unconjugated | |
2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike | |
A synthetic peptide corresponding to a sequence within amino acids 450-550 of coronavirus SARS-CoV-2 Spike RBD (N501Y) (YP_009724390.1). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction