Learn More
Invitrogen™ SARS-CoV-2 Spike Protein RBD Recombinant Llama Monoclonal Antibody (1C7)

Description
SARS-CoV-2 Spike Protein RBD Recombinant Monoclonal Antibody for ELISA, Neu
Specifications
Specifications
| Antigen | SARS-CoV-2 Spike Protein RBD |
| Applications | ELISA, Neutralization |
| Classification | Recombinant Monoclonal |
| Clone | 1C7 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Host Species | Llama |
| Immunogen | SARS-CoV-2 S protein RBD containing C-terminal His Tag. The protein was expressed in human 293 cells (HEK293). It contains amino acids Arg 319-Lys 537. |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.