Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein S1 (aa11-682), hFc-His Tag Recombinant Protein

Catalog No. PIRP87679
Encompass
Click to view available options
Quantity:
100 μg

Recombinant Protein

Reconstitution: Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water. After reconstitution, the protein solution is stable at -20°C for 3 months, at 2-8°C for up to 1 week. Description: Recombinant 2019-nCoV Spike S1 Protein is produced by HEK293 cells expression system.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, ELISA, Functional Assay, Surface Plasmon Resonance, Western Blot
Formulation PBS with no preservative; pH 7.4
Molecular Weight (g/mol) 103 kDa
Name SARS-CoV-2 Spike Protein S1 (aa11-682), hFc-His Tag
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein S1
Biological Activity Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human ACE2 (at 2 μg/mL (100 μL/well) can bind Recombinant SARS-CoV-2 Spike S1, the ED50 of SARS-COV-2 Spike S1 is 6.79 ng/mL Immobilized Human ACE2 on COOH Chip, can bind SARS-COV-2 Spike S1 with an affinity constant of 90.8 nM as determined in a SPR assay (Nicoya OpenSPR).)
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag, His-tag
Sequence SARS-CoV-2 Spike Protein S1, amino acids Val11-Arg682 (Accession # YP_009724390.1) with a C-terminal human Fc-tag and His-tag
Content And Storage -20°Cor to +80°C if preferred
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.