Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein S1/S2 (B.1.1.7) His-tag, trimer Recombinant Protein

Catalog No. PIRP88443
Encompass
Click to view available options
Quantity:
100 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV. SARS-CoV-2 spike S1/S2 (Accession # YP_009724390.1) with (69-70, 145) deletions and N501Y, A570D, D614G, T716I, F817P, A892P, A899P, A942P, S982A, K986P, V987P, D1118H substitutions and furin cleavage site mutations expressed with the bacteriophage T4 fibritin and a C-terminal His-tag. The mutations were identified in the SARS-CoV-2 varient known as B.1.1.7.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Format Lyophilized
Formulation PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4
Molecular Weight (g/mol) 136.18 kDa
Name SARS-CoV-2 Spike Protein S1/S2 (B0.1.1.7) His-tag, trimer
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
Common Name SARS-CoV-2 Spike (trimer) Protein
Biological Activity Immobilized human ACE2 protein (mFc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 B0.1.1.7 Spike S1+S2 trimer Protein (ECD, His tag) ,the EC50 of SARS-CoV-2 B0.1.1.7 Spike S1+S2 trimer Protein (ECD, His tag) is 5-30 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SARS-CoV-2 spike S1/S2 (Accession # YP_009724390.1) with variant B0.1.1.7 mutations expressed with the bacteriophage T4 fibritin and a C-terminal His-tag. Full sequence information available in Product Specfic Information section.
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized human ACE2 protein (mFc tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 B0.1.1.7 Spike S1+S2 trimer Protein (ECD, His tag) ,the EC50 of SARS-CoV-2 B0.1.1.7 Spike S1+S2 trimer Protein (ECD, His tag) is 5-30 ng/mL)
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.