Learn More
Novus Biologicals™ SARS-CoV-2 Spike RBD Antibody (Mouse IgG) - Omicron Variant, B.1.1.529 ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum |
| For Use With (Application) | Antibody Titer Assay |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | SARS-CoV-2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.