Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $351.00
Specifications
Antigen | Spike RBD |
---|---|
Clone | 1049132 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11006SP
![]() |
R&D Systems
MAB11006SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11006100
![]() |
R&D Systems
MAB11006100 |
100 μg |
Each for $351.00
|
|
|||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for ImmunocytochemistrySpecifications
Spike RBD | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Human embryonic kidney cell HEK293-derived SARS2-P.1.S, 16-1211, Accession # YP_009724390 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1049132 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
SARS-CoV-2 | |
YP_009724390 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title