Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | SARS-CoV-2 Spike RBD |
---|---|
Clone | 1034522 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10540SP
![]() |
R&D Systems
MAB10540SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10540100
![]() |
R&D Systems
MAB10540100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot.Specifications
SARS-CoV-2 Spike RBD | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
Recombinant SARS-CoV-2 Spike Protein, S1 Subunit, Receptor Binding Domain (RBD), Arg319-Phe541, Accession # YP_009724390 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1034522 | |
Western Blot | |
Unconjugated | |
Mouse | |
SARS-CoV-2 | |
YP_009724390 | |
43740568 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title