Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
| Antigen | SARS-CoV-2 Spike RBD |
|---|---|
| Clone | 1035753 |
| Dilution | ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL |
| Applications | ELISA, Blocking Assay |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB105808SP
![]() |
R&D Systems
MAB105808SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10580810
![]() |
R&D Systems
MAB105808100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for ELISA, Blockade of Receptor-ligand Interaction.Specifications
| SARS-CoV-2 Spike RBD | |
| ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived SARS-CoV-2 Spike S1 Subunit, Val16-Pro681, Accession # YP_009724390.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1035753 | |
| ELISA, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| SARS-CoV-2 | |
| YP_009724390.1 | |
| 43740568 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title