Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals SARS-CoV-2 Spike Recombinant Protein
SDP

Catalog No. NB158993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158993 100 μg
NB158994 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB158993 Supplier Novus Biologicals Supplier No. NBP314666100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Concentration 2.1 mg/ml
For Use With (Application) ELISA
Formulation 20 mM Tris, 500 mM NaCl (pH 7.5)
Gene ID (Entrez) 43740568
Quantity 100 μg
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Gene Symbol S
Research Category Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Protein SARS-CoV-2 Spike
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.