Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike S1 Antibody (B38) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP307031
Description
SARS-CoV-2 Spike S1 Monoclonal specifically detects SARS-CoV-2 Spike S1 in SARS-CoV-2 samples. It is validated for ELISA.Specifications
SARS-CoV-2 Spike S1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.5 | |
Human | |
>95% | |
RUO | |
43740568 | |
Store at 4°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
H4 | |
Western Blot, ELISA 1:5000-1:10000 | |
SARS-CoV-2 | |
Recombinant SARS-CoV-2 Spike S1 protein | |
0.05 mg | |
Primary | |
SARS-CoV-2 | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction