Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike S1 Chimeric Recombinant Human Monoclonal Antibody (hIgA2001), GenScript™

Catalog No. 502120773
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0773 Supplier Genscript Corporation Supplier No. A02071100
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.SARS-CoV-2 Spike S1 Antibody (hIgA2001), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.

SARS-CoV-2 Spike S1 Chimeric Recombinant Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Chimeric in Virus samples. It is validated for ELISA

Specifications

Antigen SARS-CoV-2 Spike S1 Chimeric
Applications ELISA
Classification Recombinant Monoclonal
Clone hIgA2001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Host Species Human
Immunogen Recombinant SARS-CoV-2 RBD Protein
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.