Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike S1 Protein Antibody (1035226), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB105805PEATT594
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Flow CytometrySpecifications
SARS-CoV-2 Spike S1 Protein | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
SARS-CoV-2 | |
Purified |
Flow Cytometry | |
1035226 | |
Flow Cytometry | |
SARS-CoV-2 | |
HEK293-derived SARS-CoV-2 Spike S1 Subunit, Accession # YP_009724390.1 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction