Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike S1 Subunit Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB105403S1
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB105403S1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB105403S1 Supplier R&D Systems Supplier No. FAB105403S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike S1 Protein Monoclonal specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Flow Cytometry.

Specifications

Antigen SARS-CoV-2 Spike S1 Protein
Applications Flow Cytometry
Classification Monoclonal
Clone 1035206
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. YP_009724390.1
Gene Alias SARS-CoV-2
Host Species Mouse
Immunogen HEK293-derived SARS-CoV-2 Spike S1 Subunit protein, Accession # YP_009724390.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.