Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike S1 Subunit Antibody, R&D Systems™

Goat nan Antibody
$200.50 - $468.00
Specifications
Antigen | SARS-CoV-2 Spike S1 Protein |
---|---|
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF10770SP
![]() |
R&D Systems
AF10770SP |
25 μg |
Each for $200.50
|
|
|||||
AF10770100
![]() |
R&D Systems
AF10770100 |
100 μg |
Each for $468.00
|
|
|||||
Description
SARS-CoV-2 Spike S1 Protein nan antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
SARS-CoV-2 Spike S1 Protein | |
Western Blot | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
SARS-CoV-2 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Western Blot 1 ug/mL | |
Unconjugated | |
Goat | |
SARS-CoV-2 | |
YP_009724390.1 | |
Chinese Hamster Ovary cell line CHO-derived SARS-CoV-2 Spike S1 Subunit, Arg319-Phe541, Accession # YP_009724390.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title