Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike S1 Subunit Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB105405SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB105405SP 25 μg
MAB10540510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB105405SP Supplier R&D Systems Supplier No. MAB105405SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike S1 Protein Monoclonal specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen SARS-CoV-2 Spike S1 Protein
Applications Blocking Assay
Classification Monoclonal
Clone 1035224
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction 50 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. YP_009724390.1
Gene Alias SARS-CoV-2
Host Species Mouse
Immunogen HEK293-derived SARS-CoV-2 Spike S1 Subunit, Accession # YP_009724390.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species SARS-CoV-2
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.