Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10889100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10889100 100 μg
MAB10889SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10889100 Supplier R&D Systems Supplier No. MAB10889100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen SARS-CoV-2 Spike S2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2824A
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_009724390.1
Gene Alias COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human SARS-CoV-2 Spike 2 protein, Met1-Lys1211, Accession # YP_009724390.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species SARS-CoV-2
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.