Learn More
Invitrogen™ SARS-CoV-2 Spike (trimer) Protein Recombinant Llama Monoclonal Antibody (T3P1D2)
Llama Recombinant Monoclonal Antibody
Supplier:  Invitrogen™ MA549350
Catalog No. PIMA549350
Only null left
Add to Cart
Description
Description
SARS-CoV-2 Spike (trimer) Protein Recombinant Monoclonal Antibody for ELISA
It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Spike (trimer) Protein
Recombinant Monoclonal
Unconjugated
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
SARS-CoV-2 variant Spike proteins.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
T3P1D2
PBS with no preservative
Llama
Affinity chromatography
RUO
Virus
Antibody
scFv
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.