Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars-Cov Nonstructural Protein 13Rabbit anti-Virus, Polyclonal, Rockland™
SDP

Catalog No. 501990109
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0109 Supplier Rockland Immunochemicals Supplier No. 100401A54
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against SARS-Coronavirus nsp13 protein. Cross reactivity with homologues from other sources has not been determined.

The coronavirus nonstructural protein 13 (nsp13) is one of the SARS-Coronavirus replicase cleaving products encoded by ORF1b. Nsp13 is thought to be part of the viral replication complex, which is associated with intracellular membranes. Nsp13 contains a C-terminal NTPase/Helicase domain and an N-terminal putative zinc-binding motif.

Specifications

Antigen Sars-Cov Nonstructural Protein 13
Applications Immunofluorescence, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.01% sodium azide; pH
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a BSA-coupled synthetic peptide corresponding to the C-terminus (amino acid residues 584-601) of the SARS Coronavirus nonstructural protein 13.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.