Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV NSP8 Antibody (5A10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313528
This item is not returnable.
View return policy
Description
SARS-CoV NSP8 Monoclonal antibody specifically detects SARS-CoV NSP8 in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| SARS-CoV NSP8 | |
| Monoclonal | |
| Unconjugated | |
| COVID-19 | |
| The immunogen used to generate this antibody corresponds to SARS virus nsp8. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| 5A10 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| SARS-CoV, SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction