Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 NSP8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $437.18
Specifications
| Antigen | SARS-CoV NSP8 |
|---|---|
| Clone | 1044736 |
| Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12502SP
![]() |
R&D Systems
MAB12502SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12502100
![]() |
R&D Systems
MAB12502100 |
100 μg |
Each for $437.18
|
|
|||||
Description
SARS-CoV NSP8 Monoclonal antibody specifically detects SARS-CoV NSP8 in SARS-CoV-2 samples. It is validated for Western Blot, Western BlotSpecifications
| SARS-CoV NSP8 | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| COVID-19 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1044736 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| SARS-CoV-2 | |
| YP_009725304.1 | |
| E. coli-derived SARS-CoV-2 NSP8 protein, Ala1-Gln198, Accession # YP_009725304.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title