Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV/SARS-CoV-2 NSP8 Monoclonal Antibody (5A10)
GREENER_CHOICE

Catalog No. PIMA535947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535947 Supplier Invitrogen™ Supplier No. MA535947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV/SARS-CoV-2 NSP8 Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), Flow

The nonstructural protein 8 (nsp8) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp8 is thought to be part of the viral replication complex, which is associated with intracellular membranes. No specific information on the function of nsp8 is available.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV/SARS-CoV-2 NSP8
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5A10
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene orf1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19; COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols orf1ab
Host Species Mouse
Immunogen SARS virus nsp8.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.