Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV/SARS-CoV-2 ORF7a Monoclonal Antibody (3C9)
GREENER_CHOICE

Catalog No. PIMA535944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535944 Supplier Invitrogen™ Supplier No. MA535944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV/SARS-CoV-2 ORF7a Monoclonal Antibody for Western Blot, ICC/IF

ORF7a is a SARS-CoV accessory protein that is composed of a type I transmembrane protein that localizes primarily to the Golgi apparatus. It can also be found on the cell surface. Its exact role in pathogenesis and disease is unknown. However, it has been observed to bind to BST-2 and inhibit BST-2 glycosylation, resulting in the subsequent loss of BST-2 antiviral function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV/SARS-CoV-2 ORF7a
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ORF7a
Gene Accession No. 0
Gene Alias 2019-nCoV orf7a; Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 orf7a; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV orf7a; SARS-CoV-2; SARS-CoV-2 orf7a
Gene Symbols ORF7a
Host Species Mouse
Immunogen SARS-CoV ORF7a protein (16-111 a.a.).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740573
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.